Learn More
(2S,3R)-3-Hydroxynorleucine, TRC
Product Code. 30402121
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. Quantity Unit Size
30402121 50 mg 50mg
1 options
Product Code. 30402121 Supplier Toronto Research Chemicals Supplier No. TRCH94976050MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2S,3R)-3-Hydroxynorleucine
CAS 10148-69-3
Molecular Formula C6H13NO3
Quantity 50 mg
InChI Formula InChI=1S/C6H13NO3/c1-2-3-4(8)5(7)6(9)10/h4-5,8H,2-3,7H2,1H3,(H,9,10)/t4-,5+/m1/s1
SMILES N[C@H](C(O)=O)[C@H](O)CCC
IUPAC Name (2S,3R)-2-amino-3-hydroxyhexanoic acid
Molecular Weight (g/mol) 147.17
Formula Weight 147.0895