Läs mer
(2S,3S)-3-(2-Aminophenyl)sulfanyl-2-hydroxy-3-(4-methoxyphenyl)propanoic Acid, Mikromol™
Produktkod. 30460793
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30460793 4 x 25 mg
30052078 25 mg
30052052 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30460793 Leverantör Mikromol Leverantörsnummer MM0068.1300254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2S,3S)-3-(2-Aminophenyl)sulfanyl-2-hydroxy-3-(4-methoxyphenyl)propanoic Acid, Mikromol
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2S,3S)-3-(2-Aminophenyl)sulfanyl-2-hydroxy-3-(4-methoxyphenyl)propanoic Acid
CAS 42399-48-4
Analyt- eller komponentnamn (2S,3S)-3-(2-Aminophenyl)sulfanyl-2-hydroxy-3-(4-methoxyphenyl)propanoic Acid
Frakt skick Room Temperature
Molekylformel C16 H17 N O4 S
Certifikat och efterlevnad ISO 17025
Synonym (2S,3S)-3-(2-Aminophenyl)sulfanyl-2-hydroxy-3-(4-methoxyphenyl)propanoic Acid, (alphaS,betaS)-beta-[(2-Aminophenyl)thio]-alpha-hydroxy-4-methoxybenzenepropanoic acid, Benzenepropanoic acid, beta-[(2-aminophenyl)thio]-alpha-hydroxy-4-methoxy-, [S-(R*,R*)]-, d-threo-2-Hydroxy-3-(2-aminophenylthio)-3-(4-methoxyphenyl)propionic acid
InChI formel InChI=1S/C16H17NO4S/c1-21-11-8-6-10(7-9-11)15(14(18)16(19)20)22-13-5-3-2-4-12(13)17/h2-9,14-15,18H,17H2,1H3,(H,19,20)/t14-,15+/m1/s1
Lösningstyp Neat
LEDER COc1ccc(cc1)[C@H](Sc2ccccc2N)[C@@H](O)C(=O)O
IUPAC-namn (2S,3S)-3-(2-aminophenyl)sulfanyl-2-hydroxy-3-(4-methoxyphenyl)propanoic acid
Molekylvikt (g/mol) 319.38
Kvantitet 4 x 25 mg
Formel vikt 319.0878 g/mol