Läs mer
(2S,3S)-3-Hydroxy-2-(4-methoxyphenyl)-2,3-dihydro-1,5-benzothiazepin-4(5H)-one, LoGiCal
Produktkod. 30464193
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30464193 10 mg
1 options
Produktkod. 30464193 Leverantör LoGiCal Leverantörsnummer LGCFOR0068.04
LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
(2S,3S)-3-Hydroxy-2-(4-methoxyphenyl)-2,3-dihydro-1,5-benzothiazepin-4(5H)-one
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2 S,3 S)-3-Hydroxy-2-(4-methoxyphenyl)-2,3-dihydro-1,5-benzothiazepin-4(5 H)-one
CAS 42399-49-5
Analyt- eller komponentnamn (2 S,3 S)-3-Hydroxy-2-(4-methoxyphenyl)-2,3-dihydro-1,5-benzothiazepin-4(5 H)-one
Frakt skick Room Temperature
Molekylformel C16 H15 N O3 S
Certifikat och efterlevnad ISO 9001
Synonym Diltiazem Hydrochloride Imp. E (EP), Diltiazem Imp. E (EP), (2 S,3 S)-3-Hydroxy-2-(4-methoxyphenyl)-2,3-dihydro-1,5-benzothiazepin-4(5 H)-one, Diltiazem Hydrochloride Impurity E, Diltiazem Impurity E
InChI formel InChI=1 S/C16H15NO3S/c1-20-11-8-6-10(7-9-11)15-14(18)16(19)17-12-4-2-3-5-13(12)21-15/h2-9,14-15,18 H,1H3,(H,17,19)/t14-,15+/m1/s1
LEDER COc1ccc(cc1)[C@@H]2 Sc3ccccc3NC(=O)[C@@H]2 O
IUPAC-namn (2 S,3 S)-3-hydroxy-2-(4-methoxyphenyl)-3,5-dihydro-2 H-1,5-benzothiazepin-4-one
Industry Type Forensics
Molekylvikt (g/mol) 301.36
Kvantitet 10 mg
Formel vikt 301.0773 g/mol