Läs mer
(2S,3S)-3-Hydroxy-2-methylbutanoic Sodium Salt, TRC
Produktkod. 30408049
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30408049 50 mg
30407882 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30408049 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH94664050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2S,3S)-3-Hydroxy-2-methylbutanoic Sodium Salt
CAS 66767-64-4
Molekylformel C5H9NaO3
Kvantitet 50 mg
InChI formel InChI=1S/C5H10O3.Na/c1-3(4(2)6)5(7)8;/h3-4,6H,1-2H3,(H,7,8);/q;+1/p-1
LEDER O[C@H]([C@H](C)C(O[Na])=O)C
IUPAC-namn sodium (2S,3S)-3-hydroxy-2-methylbutanoate
Molekylvikt (g/mol) 140.11
Formel vikt 140.0449