Läs mer
(2S,3S,4aR,6S,8R,8aR)-Octahydro-6,8-dihydroxy-2,3-dimethoxy-2,3-dimethyl-1,4-benzodioxin-6-carboxylic Acid Methyl Ester, TRC
Produktkod. 30413120
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30413120 250 mg
1 options
Produktkod. 30413120 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO236350250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2S,3S,4aR,6S,8R,8aR)-Octahydro-6,8-dihydroxy-2,3-dimethoxy-2,3-dimethyl-1,4-benzodioxin-6-carboxylic Acid Methyl Ester
CAS 176798-26-8
Molekylformel C14H24O8
Kvantitet 250 mg
Synonym [2S-(2alpha,3beta,4abeta,6beta,8beta,8aalpha)]-Octahydro-6,8-dihydroxy-2,3-dimethoxy-2,3-dimethyl-1,4-benzodioxin-6-carboxylic Acid Methyl Ester
InChI formel InChI=1S/C14H24O8/c1-12(19-4)13(2,20-5)22-10-8(15)6-14(17,11(16)18-3)7-9(10)21-12/h8-10,15,17H,6-7H2,1-5H3/t8-,9-,10-,12+,13+,14+/m1/s1
LEDER COC(=O)[C@]1(O)C[C@@H](O)[C@H]2O[C@](C)(OC)[C@@](C)(OC)O[C@@H]2C1
IUPAC-namn methyl (2S,3S,4aR,6S,8R,8aR)-6,8-dihydroxy-2,3-dimethoxy-2,3-dimethyl-5,7,8,8a-tetrahydro-4aH-benzo[b][1,4]dioxine-6-carboxylate
Molekylvikt (g/mol) 320.34
Formel vikt 320.1471