Läs mer
(2S)-4-Oxo-2-(3-thiazolidinylcarbonyl)-1-pyrrolidinecarboxylic Acid 1,1-Dimethylethyl Ester, TRC
Produktkod. 30419435
Change view
Kvantitet:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30419435 500 mg
30408942 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30419435 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO870490500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 401564-36-1
Molekylformel C13H20N2O4S
Molekylvikt (g/mol) 300.37
IUPAC-namn tert-butyl (S)-4-oxo-2-(thiazolidine-3-carbonyl)pyrrolidine-1-carboxylate
LEDER CC(C)(C)OC(N1[C@@H](CC(C1)=O)C(N2CCSC2)=O)=O
Specifikationer
Kemiskt namn eller material (2S)-4-Oxo-2-(3-thiazolidinylcarbonyl)-1-pyrrolidinecarboxylic Acid 1,1-Dimethylethyl Ester
Kvantitet 500 mg
InChI formel InChI=1S/C13H20N2O4S/c1-13(2,3)19-12(18)15-7-9(16)6-10(15)11(17)14-4-5-20-8-14/h10H,4-8H2,1-3H3/t10-/m0/s1
Formel vikt 300.11