Läs mer
(2S,4R)-Itraconazole (Mixture of Diastereomers), TRC
Produktkod. 30406216
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30406216 1 mg
1 options
Produktkod. 30406216 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI9375351MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2S,4R)-Itraconazole (Mixture of Diastereomers)
CAS 939027-87-9
Molekylformel C35H38Cl2N8O4
Kvantitet 1 mg
Synonym 4-[4-[4-[4-[[(2S,4R)-2-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]-1-piperazinyl]phenyl]-2,4-dihydro-2-(1-methylpropyl)-3H-1,2,4-triazol-3-one
InChI formel InChI=1S/C35H38Cl2N8O4/c1-3-25(2)45-34(46)44(24-40-45)29-7-5-27(6-8-29)41-14-16-42(17-15-41)28-9-11-30(12-10-28)47-19-31-20-48-35(49-31,21-43-23-38-22-39-43)32-13-4-26(36)18-33(32)37/h4-13,18,22-25,31H,3,14-17,19-21H2,1-2H3/t25?,31-,35-/m1/s1
LEDER O=C1N(C(C)CC)N=CN1C(C=C2)=CC=C2N(CC3)CCN3C4=CC=C(OC[C@H]5O[C@](CN6N=CN=C6)(C7=CC=C(Cl)C=C7Cl)OC5)C=C4
Molekylvikt (g/mol) 705.63