Läs mer
(2S,4S)-4-Cyclohexyl-1-(3-oxopentanoyl)pyrrolidine-2-carboxylic Acid, TRC
Produktkod. 30354843
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30354843 50 mg
1 options
Produktkod. 30354843 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC98456150MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Fosinopril Impurity H
CAS 149760-14-5
Kvantitet 50 mg
Synonym trans-4-cyclohexyl-1-(1,3-dioxopentyl)-L-Proline (9CI), L-Proline, 4-cyclohexyl-1-(1,3-dioxopentyl)-, trans- (9CI)
LEDER CCC(=O)CC(=O)N1C[C@@H](C[C@H]1C(=O)O)C2CCCCC2