Läs mer
(2S,4S)-Sacubitril, TRC
Produktkod. 30009800
Change view
Kvantitet:
2.5 mg
5 mg
10 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30009800 2.5 mg 2.5mg
30004655 5 mg -
30009161 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009800 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS0809152.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 149709-63-7
Molekylformel C24 H29 N O5
Molekylvikt (g/mol) 411.49
Synonym [S-(R*,R*)]-γ-[(3-Carboxy-1-oxopropyl)amino]-α-methyl-[1,1’-biphenyl]-4-pentanoic Acid Ethyl Ester, Sacubitril-(2S,4S) Isomer, 4-(((2S,4S)-1-([1,1’-Biphenyl]-4-yl)-5-ethoxy-4-methyl-5-oxopentan-2-yl)amino)-4-oxobutanoic Acid;4-(((2S,4S)-1-([1,1'-Biphenyl]-4-yl)-5-ethoxy-4-methyl-5-oxopentan-2-yl)amino)-4-oxobutanoic acid;(4-(((2S,4S)-1-([1,1'-Biphenyl]-4-yl)-5-ethoxy-4-methyl-5-oxopentan-2-yl)amino)-4-oxobutanoic acid)
IUPAC-namn (4Z)-4-[(1S)-4-ethoxy-3-methyl-4-oxo-1-[(4-phenylphenyl)methyl]butyl]imino-4-hydroxy-butanoic acid
LEDER CCOC(=O)C(C)C[C@@H](Cc1ccc(cc1)c2ccccc2)\N=C(/O)\CCC(=O)O
Specifikationer
Kemiskt namn eller material (2S,4S)-Sacubitril
Kvantitet 2.5 mg
InChI formel InChI=1S/C24H29NO5/c1-3-30-24(29)17(2)15-21(25-22(26)13-14-23(27)28)16-18-9-11-20(12-10-18)19-7-5-4-6-8-19/h4-12,17,21H,3,13-16H2,1-2H3,(H,25,26)(H,27,28)/t17?,21-/m0/s1
Formel vikt 411.2046