Läs mer
(2S,5R,6R)-3,3-Dimethyl-7-oxo-6-[[2-(4-hydroxyphenoxy)acetyl]amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid (4-Hydroxyphenoxymethylpenicillin), Mikromol™
Produktkod. 30050636
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30050636 100 mg
30052272 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30050636 Leverantör Mikromol Leverantörsnummer MM0026.02
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2S,5R,6R)-3,3-Dimethyl-7-oxo-6-[[2-(4-hydroxyphenoxy)acetyl]amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid (4-Hydroxyphenoxymethylpenicillin)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Hydroxyphenoxymethylpenicillin
CAS 20880-67-5
Analyt- eller komponentnamn 4-Hydroxyphenoxymethylpenicillin
Frakt skick Ice pack (-20°C)
Molekylformel C16 H18 N2 O6 S
Certifikat och efterlevnad ISO 9001
Synonym Phenoxymethylpenicillin Potassium Imp. D (EP), Phenoxymethylpenicillin Imp. D (EP), (2S,5R,6R)-3,3-Dimethyl-7-oxo-6-[[2-(4-hydroxyphenoxy)acetyl]amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylicAcid, 4-Hydroxyphenoxymethylpenicillin, Phenoxymethylpenicillin Potassium Impurity D, Phenoxymethylpenicillin Impurity D
InChI formel InChI=1S/C16H18N2O6S/c1-16(2)12(15(22)23)18-13(21)11(14(18)25-16)17-10(20)7-24-9-5-3-8(19)4-6-9/h3-6,11-12,14,19H,7H2,1-2H3,(H,17,20)(H,22,23)/t11-,12+,14-/m1/s1
LEDER CC1(C)S[C@@H]2[C@H](NC(=O)COc3ccc(O)cc3)C(=O)N2[C@H]1C(=O)O
IUPAC-namn (2S,5R,6R)-6-[[2-(4-hydroxyphenoxy)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Molekylvikt (g/mol) 366.39
Kvantitet 100 mg
Formel vikt 366.0886 g/mol