missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (2S,5S)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-diethylphospholane, 97%

Brand:  Alfa Aesar™ H63109.MC

Additional Details : CAS Number : 1314246-02-0 Weight : 0.00010kg

Quantity 100mg

 View more versions of this product

Product Code. 15478126

  • 726.00 SEK / 100mg


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

(2S,5S)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-diethylphospholane
292.35
S,S-Et-RajPhos
1314246-02-0
MFCD09265099

Specifications

(2S,5S)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-diethylphospholane
100mg
1.076g/mL
97%
MFCD09265099
1.5580
Air Sensitive
1314246-02-0
Liquid
292.35
S,S-Et-RajPhos
Pale yellow to colorless
Videos
SDS