Läs mer
Tocris Bioscience™ 2S,6S-Hydroxynorketamine hydrochloride
Beskrivning
2S,6S-Hydroxynorketamine hydrochloride is a ketamine metabolite. Decreases intracellular DL>D-serine (a NMDA co-agonist) concentrations in PC-12 cells (IC50 = 0.18 nM). Exerts antidepressant effects in mice. 2R,6R-Enantiomer and Racemate also available.
Specifikationer
Specifikationer
| Kemiskt namn eller material | (2S,6S)-2-Amino-2-(2-chlorophenyl)-6-hydroxycyclohexanone hydrochloride |
| CAS | 1430202-70-2 |
| Kvantitet | 10 mg |
| Mål | AMPA Receptor Agonists |
| Molekylformel | C12H14ClNO2.HCl |
| Renhet | 98% |