Läs mer
(2S)-Ethylpyrrolidine Hydrochloride, TRC
Produktkod. 30373064
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30373064 100 mg
1 options
Produktkod. 30373064 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE818080100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2S)-Ethylpyrrolidine Hydrochloride
CAS 597562-96-4
Molekylformel C6H14ClN
Kvantitet 100 mg
Synonym (2S)-2-Ethylpyrrolidine Hydrochloride, (2S)-2-Ethylpyrrolidinium Chloride
InChI formel InChI=1S/C6H13N.ClH/c1-2-6-4-3-5-7-6;/h6-7H,2-5H2,1H3;1H/t6-;/m0./s1
LEDER OC[C@@H]1C(O)C[C@@H](O1)[n+]2c3c(C(=O)NC([NH-])=N3)n(CC)c2
Molekylvikt (g/mol) 135.64