Läs mer
(2S)-pentan-2-amine hydrochloride, TRC
Produktkod. 30419578
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30419578 100 mg
1 options
Produktkod. 30419578 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS681758100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2S)-2-Pentanamine Hydrochloride
CAS 216237-52-4
Molekylformel C5H13N • HCl
Kvantitet 100 mg
Synonym 2-Pentanamine','hydrochloride (1:1)','(2S)- (ACI), 2-Pentanamine','hydrochloride','(2S)- (9CI), (S)-Pentan-2-amine hydrochloride, (2S)-2-Pentanamine hydrochloride, (S)-2-Aminopentane hydrochloride, (S)-1-Methylbutylamine hydrochloride, (S)-2-Pentylamine hydrochloride
InChI formel InChI=1S/C5H13N.ClH/c1-3-4-5(2)6;/h5H,3-4,6H2,1-2H3;1H/t5-;/m0./s1
LEDER CCC[C@@H](N)C.Cl
IUPAC-namn (2S)-pentan-2-amine;hydrochloride
Molekylvikt (g/mol) 87.163646
Formel vikt 123.0815