missing translation for 'onlineSavingsMsg'
Läs mer

Gibco™ Human Granzyme B Recombinant Protein, PeproTech®

Produktkod. 30019313
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
10 μg
100 μg
2 μg
250 μg
500 μg
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 30019313

Brand: Gibco™ 140182UG

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Recombinant Protein

Recombinant Human Granzyme B has been confirmed to be a mixture of the pro-peptide and mature forms when tested by N-terminal sequencing. Recombinant Human Granzyme B is a 235 amino acid protein, which includes the mature 227 amino acid sequence, as well as an 8 amino acid C-terminal His-tag. Due to glycosylation, this protein migrates to an approximate molecular weight of 30-40 kDa, under reducing and non-reducing conditions.

Granzyme B is a member of the granzyme serine protease family, and is found in the granules of cytotoxic T cells and NK cells. Granzyme B has been described as CGL1 (cathepsin G-like-1), a serine protease expressed only in cytotoxic T-lymphocytes after cell activation, and CTLA-1 (cytotoxic T lymphocyte-associated serine esterase 1) based on identification of mRNA in various cytotoxic T cells, but not observed in non-cytotoxic lymphoid cells. Granzyme B is crucial for the rapid induction of target cell death by apoptosis, induced by interaction with cytotoxic T cells. The receptor involved in this process has been identified as mannose 6-phosphate receptor which functions as a death receptor for Granzyme B during cytotoxic T cell-induced apoptosis. Granzyme B enters target cells to cleave caspase-3 and initiate the caspase cascade leading to DNA fragmentation and apoptosis. Granzyme B can also act through a mitochondrial apoptosis pathway by cleaving the Bid protein. Granzymes are neutral serine proteases, which are stored in specialized lytic granules of cytotoxic T lymphocytes (CTLs) and in natural killer (NK) cells. A number of granzymes (A to G) have been isolated and cloned from mouse CTLs and NK cells, however in man, fewer have been cloned and identified.
TRUSTED_SUSTAINABILITY

Specifikationer

Tillträdesnummer P10144
För användning med (applikation) Functional Assay
Formatera Lyophilized
Formulering protein with no preservative
Gen-ID (Entrez) 3002
Molekylvikt (g/mol) 26.6 kDa
Namn Human Granzyme B
Kvantitet 2 μg
Regulatorisk status RUO
Endotoxinkoncentration <1 EU/ μg
Gene Alias AI553453; C11; Cathepsin G-like 1; CCP1; CCP-1/C11; CCPI; CGL1; CGL-1; CSPB; CSP-B; Ctla1; Ctla-1; CTSGL1; cytotoxic cell protease 1; cytotoxic serine protease B; Cytotoxic T lymphocyte associated serine esterase 1; cytotoxic T-lymphocyte proteinase 2; cytotoxic T-lymphocyte-associated serine esterase 1; fragmentin; fragmentin 2; fragmentin-2; GLP I; GLP III; GLP-1; granzyme 2; granzyme B; granzyme B (granzyme 2, cytotoxic T-lymphocyte-associated serine esterase 1); granzyme B(G,H); Granzyme-2; GranzymeB; granzyme-like protein 1; granzyme-like protein I; GRB; GZB; Gzmb; HLP; human lymphocyte protein; Human lymphocyte protein (Hlp); Lymphocyte protease; Natural killer cell protease 1; OTTHUMP00000028189; RNKP-1; SECT; T-cell serine protease 1-3 E
Vanligt namn Granzyme B
Biologisk aktivitet Determined by its ability to cleave chromogentic Granzyme B substrate IEPD-pNA
Konjugera Unconjugated
Rekombinant Recombinant
Sekvens IIGGHEAKPH SRPYMAYLMI WDQKSLKRCG GFLIRDDFVL TAAHCWGSSI NVTLGAHNIK EQEPTQQFIP VKRPIPHPAY NPKNFSNDIM LLQLERKAKR TRAVQPLRLP SNKAQVKPGQ TCSVAGWGQT APLGKHSHTL QEVKMTVQED RKCESDLRHY YDSTIELCVG DPEIKKTSFK GDSGGPLVCN KVAQGIVSYG RNNGMPPRAC TKVSSFVHWI KKTMKRYHHH HHHHH
Innehåll och lagring -20°C
Uttryckssystem CHO cells
Form Lyophilized
Renhet eller kvalitetsklass ≥ 95% by SDS-PAGE gel and HPLC analyses
Visa mer Visa mindre
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.