Läs mer
(2Z)-3-Methyl-4-(benzyloxy)-2-buten-1-ol 1-Phosphate Dimethyl Diester, TRC
Produktkod. 30476967
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30476967 100 mg
1 options
Produktkod. 30476967 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM292710100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2Z)-3-Methyl-4-(benzyloxy)-2-buten-1-ol 1-Phosphate Dimethyl Diester
CAS 1798428-35-9
Molekylformel C14H21O5P
Kvantitet 100 mg
Synonym (Z)-4-Benzyloxy-3-methylbut-2-en-1-ol 1-Phosphate Dimethyl Diester
InChI formel InChI=1S/C14H21O5P/c1-13(9-10-19-20(15,16-2)17-3)11-18-12-14-7-5-4-6-8-14/h4-9H,10-12H2,1-3H3/b13-9-
LEDER C/C(COCC1=CC=CC=C1)=C/COP(OC)(OC)=O
IUPAC-namn (Z)-4-(benzyloxy)-3-methylbut-2-en-1-yl dimethyl phosphate
Molekylvikt (g/mol) 300.29
Formel vikt 300.11