Läs mer
(2Z)-3-Methyl-4-(benzyloxy)-2-butenoic Acid Methyl Ester, TRC
Produktkod. 30478396
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30478396 10 mg
1 options
Produktkod. 30478396 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM28897010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2Z)-3-Methyl-4-(benzyloxy)-2-butenoic Acid Methyl Ester
CAS 449759-41-5
Molekylformel C13H16O3
Kvantitet 10 mg
Synonym (2Z)-3-Methyl-4-(phenylmethoxy)-2-butenoic Acid Methyl Ester
InChI formel InChI=1S/C13H16O3/c1-11(8-13(14)15-2)9-16-10-12-6-4-3-5-7-12/h3-8H,9-10H2,1-2H3/b11-8-
LEDER COC(/C=C(COCC1=CC=CC=C1)/C)=O
IUPAC-namn methyl (Z)-4-(benzyloxy)-3-methylbut-2-enoate
Molekylvikt (g/mol) 220.26
Formel vikt 220.11