Läs mer
3-(1,2,4-Triazol-1-yl)-L-alanine, TRC
Produktkod. 30450993
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30450993 50 mg
1 options
Produktkod. 30450993 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT76751550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-(1,2,4-Triazol-1-yl)-L-alanine
CAS 4819-36-7
Molekylformel C5 H8 N4 O2
Kvantitet 50 mg
Synonym 1H-1,2,4-Triazole-1-propanoic acid','α-amino-','(αS)- (9CI','ACI), (αS)-α-Amino-1H-1,2,4-triazole-1-propanoic acid (ACI), 1H-1,2,4-Triazole-1-propanoic acid','α-amino-','(S)- (ZCI), 1H-1,2,4-Triazole-1-propionic acid','α-amino-','L- (8CI), (2S)-2-Amino-3-(1H-1,2,4-triazol-1-yl)propanoic acid, 3-(1,2,4-Triazol-1-yl)-L-alanine
InChI formel InChI=1S/C5H8N4O2/c6-4(5(10)11)1-9-3-7-2-8-9/h2-4H,1,6H2,(H,10,11)/t4-/m0/s1
LEDER N[C@@H](Cn1cncn1)C(=O)O
IUPAC-namn (2S)-2-amino-3-(1,2,4-triazol-1-yl)propanoic acid
Molekylvikt (g/mol) 156.14
Formel vikt 156.0647