missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(1,4-Dioxaspiro[4.5]dec-7-en-8-yl)-5-nitroindole, 97%

Brand:  Alfa Aesar™ H63266.06

Additional Details : CAS Number : 915037-53-5 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15478206

  • 6488.72 SEK / 5g


Estimated Shipment date
from Supplier 02-07-2018
Add to basket

Specifications

Chemical Identifiers

3-(1,4-Dioxaspiro[4.5]dec-7-en-8-yl)-5-nitroindole
915037-53-5
300.31
MFCD28126767

Specifications

3-(1,4-Dioxaspiro[4.5]dec-7-en-8-yl)-5-nitroindole
915037-53-5
97%
MFCD28126767
5g
Solid
300.31
Videos
SDS