missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(1,4-Dioxaspiro[4.5]dec-7-en-8-yl)-6-nitroindole, 97%

Brand:  Alfa Aesar™ H63441.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15478306

  • 10374.25 SEK / 5g


Estimated Shipment date
from Supplier 06-08-2018
Add to basket

Specifications

Chemical Identifiers

3-(1,4-Dioxaspiro[4.5]dec-7-en-8-yl)-6-nitroindole
MFCD28098697
15578032

Specifications

3-(1,4-Dioxaspiro[4.5]dec-7-en-8-yl)-6-nitroindole
5g
97%
15578032
300.31
Solid
MFCD28098697
Videos
SDS