missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(1,4-Dioxaspiro[4.5]dec-7-en-8-yl)indole, 97%

Brand:  Alfa Aesar™ H63998.MD

Additional Details : CAS Number : 143165-32-6 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15478626

  • 648.87 SEK / 250mg


Estimated Shipment date
from Supplier 06-07-2018
Add to basket

Specifications

Chemical Identifiers

3-(1,4-Dioxaspiro[4.5]dec-7-en-8-yl)indole
255.31
7644105
143165-32-6
MFCD06411611

Specifications

3-(1,4-Dioxaspiro[4.5]dec-7-en-8-yl)indole
250mg
Solid
255.31
7644105
194° to 196°C
143165-32-6
97%
MFCD06411611
Videos
SDS