missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(1-Piperidinylmethyl)indole, 95%

Brand:  Alfa Aesar™ H63256.06

Additional Details : CAS Number : 5355-42-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15449858

  • 10162.13 SEK / 5g


Estimated Shipment date
from Supplier 20-11-2018
Add to basket

Description

Description

Applications
3-(1-Piperidinylmethyl)indole, is used as a pharmaceutical intermediate.

Solubility
Freely soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

5355-42-0
214.312
HPKXPFRRVVWAPD-UHFFFAOYSA-N
21454
C1CCN(CC1)CC2=CNC3=CC=CC=C32
C14H18N2
MFCD00458412
n-skatylpiperidine, 3-piperidin-1-yl methyl-1h-indole, 3-piperidinomethyl indole, indole, 3-piperidinomethyl, 3-piperidin-1-ylmethyl-1h-indole, chembl57535, 3-piperidinomethyl-1h-indole, 3-1-piperidinylmethyl-1h-indole, 3-piperidylmethyl indole, 3-piperidinomethyl indole
3-(piperidin-1-ylmethyl)-1H-indole

Specifications

3-(1-Piperidinylmethyl)indole
5g
C14H18N2
176547
HPKXPFRRVVWAPD-UHFFFAOYSA-N
3-(piperidin-1-ylmethyl)-1H-indole
21454
Solid
Tan to White
5355-42-0
MFCD00458412
n-skatylpiperidine, 3-piperidin-1-yl methyl-1h-indole, 3-piperidinomethyl indole, indole, 3-piperidinomethyl, 3-piperidin-1-ylmethyl-1h-indole, chembl57535, 3-piperidinomethyl-1h-indole, 3-1-piperidinylmethyl-1h-indole, 3-piperidylmethyl indole, 3-piperidinomethyl indole
C1CCN(CC1)CC2=CNC3=CC=CC=C32
214.312
214.3
95%
Videos
SDS