missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Piperidinylmethyl)indole, 95%, Thermo Scientific™

Catalog Number p-7054833
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
5355-42-0
C14H18N2
214.312
MFCD00458412
HPKXPFRRVVWAPD-UHFFFAOYSA-N
n-skatylpiperidine, 3-piperidin-1-yl methyl-1h-indole, 3-piperidinomethyl indole, indole, 3-piperidinomethyl, 3-piperidin-1-ylmethyl-1h-indole, chembl57535, 3-piperidinomethyl-1h-indole, 3-1-piperidinylmethyl-1h-indole, 3-piperidylmethyl indole
21454
3-(piperidin-1-ylmethyl)-1H-indole
C1CCN(CC1)CC2=CNC3=CC=CC=C32
This item is not returnable. View return policy
5355-42-0
C14H18N2
214.312
MFCD00458412
HPKXPFRRVVWAPD-UHFFFAOYSA-N
n-skatylpiperidine, 3-piperidin-1-yl methyl-1h-indole, 3-piperidinomethyl indole, indole, 3-piperidinomethyl, 3-piperidin-1-ylmethyl-1h-indole, chembl57535, 3-piperidinomethyl-1h-indole, 3-1-piperidinylmethyl-1h-indole, 3-piperidylmethyl indole
21454
3-(piperidin-1-ylmethyl)-1H-indole
C1CCN(CC1)CC2=CNC3=CC=CC=C32

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-(1-Piperidinylmethyl)indole, is used as a pharmaceutical intermediate.

Solubility
Freely soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Color Tan to White
Quantity 250 mg
Beilstein 176547
Solubility Information Freely soluble in water.
Formula Weight 214.3
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-(1-Piperidinylmethyl)indole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'