Läs mer
3,12-Diketo-5Beta-cholanoic Acid, TRC
Produktkod. 30373179
Change view
Kvantitet:
10 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30373179 10 mg
30369454 500 mg
30372692 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30373179 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD94390010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2958-05-6
Molekylformel C24H36O4
Molekylvikt (g/mol) 388.54
Synonym 3,12-Dioxodeoxycholic Acid, Dehydrodeoxycholic Acid, (5beta)-3,12-Dioxocholan-24-oic Acid, 5b-CHOLANIC ACID-3,12-DIONE
LEDER C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4CC(=O)CC[C@]4(C)[C@H]3CC(=O)[C@]12C
Specifikationer
Kemiskt namn eller material 3,12-Diketo-5beta-cholanoic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C24H36O4/c1-14(4-9-22(27)28)18-7-8-19-17-6-5-15-12-16(25)10-11-23(15,2)20(17)13-21(26)24(18,19)3/h14-15,17-20H,4-13H2,1-3H3,(H,27,28)/t14-,15-,17+,18-,19+,20+,23+,24-/m1/s1
Formel vikt 388.2614
Procent renhet >95%