Läs mer
3-(1H-Pyrazol-1-yl)alanine, TRC
Produktkod. 30424981
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30424981 500 mg
30408969 250 mg
30412691 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30424981 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP842245500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2734-48-7
Molekylformel C6H9N3O2
Molekylvikt (g/mol) 155.15
Synonym (alphaS)-alpha-Amino-1H-pyrazole-1-propanoic Acid, L-beta-(1-Pyrazolyl)alanine, beta-(Pyrazol-1-yl)-L-alanine, beta-Pyrazolyl-L-alanine
IUPAC-namn (2S)-2-amino-3-pyrazol-1-ylpropanoic acid
LEDER N[C@@H](Cn1cccn1)C(=O)O
Specifikationer
Kemiskt namn eller material 3-(1H-Pyrazol-1-yl)alanine
InChI formel InChI=1S/C6H9N3O2/c7-5(6(10)11)4-9-3-1-2-8-9/h1-3,5H,4,7H2,(H,10,11)/t5-/m0/s1
Kvantitet 500 mg
Formel vikt 155.0695