Learn More
3-(1H-Pyrrol-1-yl)aniline, 97%, Thermo Scientific™
Catalog Number p-2698860
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
89353-42-4
C10H10N2
158.204
MFCD03407309
PJGDCPOPSNUYHC-UHFFFAOYSA-N
3-1h-pyrrol-1-yl aniline, 3-pyrrol-1-yl-phenylamine, 3-pyrrol-1-yl aniline, 3-pyrrolylphenylamine, 3-1-pyrrolyl aniline, 3-pyrrol-1-yl-aniline, 1-3-aminophenyl pyrrole, benzenamine, 3-1h-pyrrol-1-yl
2760546
3-pyrrol-1-ylaniline
C1=CN(C=C1)C2=CC(=CC=C2)N
89353-42-4
C10H10N2
158.204
MFCD03407309
PJGDCPOPSNUYHC-UHFFFAOYSA-N
3-1h-pyrrol-1-yl aniline, 3-pyrrol-1-yl-phenylamine, 3-pyrrol-1-yl aniline, 3-pyrrolylphenylamine, 3-1-pyrrolyl aniline, 3-pyrrol-1-yl-aniline, 1-3-aminophenyl pyrrole, benzenamine, 3-1h-pyrrol-1-yl
2760546
3-pyrrol-1-ylaniline
C1=CN(C=C1)C2=CC(=CC=C2)N
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 158.2
Percent Purity 97%
Chemical Name or Material 3-(1H-pyrrol-1-yl)aniline