Läs mer
3-(1H-Pyrrol-2-yl)pyridine (beta-Nornicotyrine), Mikromol™
Produktkod. 30049716
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30049716 25 mg
17921956 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30049716 Leverantör Mikromol Leverantörsnummer MM0517.150025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-(1H-Pyrrol-2-yl)pyridine (beta-Nornicotyrine)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-(1H-Pyrrol-2-yl)pyridine (beta-Nornicotyrine)
CAS 494-98-4
Analyt- eller komponentnamn 3-(1H-Pyrrol-2-yl)pyridine (beta-Nornicotyrine)
Frakt skick Room Temperature
Molekylformel C9H8N2
Certifikat och efterlevnad ISO 17025
Synonym Pyridine, 3-(1H-pyrrol-2-yl)-, Pyridine, 3-pyrrol-2-yl- (6CI,7CI,8CI), Nornicotyrine, β-Nornicotyrine, 3-(1H-Pyrrol-2-yl)pyridine (Beta-Nornicotyrine), Beta-Nornicotyrine, beta-Nornicotyrine
InChI formel InChI=1S/C9H8N2/c1-3-8(7-10-5-1)9-4-2-6-11-9/h1-7,11H
LEDER c1cncc(c1)c2ccc[nH]2
IUPAC-namn 3-(1H-pyrrol-2-yl)pyridine
Molekylvikt (g/mol) 144.1732
Kvantitet 25 mg
Formel vikt 144.0687 g/mol