Learn More
3-(2,2,3-Trimethylnorborn-5-yl)cyclohexyl acrylate, Thermo Scientific Chemicals
Catalog Number p-7080141
Click to view available options
Quantity:
5 g
25 g
100 g
903876-45-9
C19H30O2
290.45
MFCD22380703
CDBRNGRSVNBVLJ-UHFFFAOYNA-N
3-5,5,6-trimethylbicyclo 2.2.1 heptan-2-yl cyclohexyl prop-2-enoate
903876-45-9
C19H30O2
290.45
MFCD22380703
CDBRNGRSVNBVLJ-UHFFFAOYNA-N
3-5,5,6-trimethylbicyclo 2.2.1 heptan-2-yl cyclohexyl prop-2-enoate
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Color Brown
Flash Point >149°C (300°F)
Quantity 5 g
Formula Weight 290.45
Physical Form Liquid
Chemical Name or Material 3- (2,2,3-Trimethylnorborn-5-yl)cyclohexyl acrylate