Läs mer
3-[[2-[(2-Chloroethyl)amino]ethyl]amino]propyl Monophosphate Dihydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 3-[2-(2-chloroethylamino)ethylamino]propyl dihydrogen phosphate dihydrochloride |
| CAS | 158401-51-5 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C7 H18 Cl N2 O4 P . 2 Cl H |
| Kvantitet | 1 g |
| Synonym | 1-Propanol, 3-[[2-[(2-chloroethyl)amino]ethyl]amino]-, dihydrogen phosphate (ester), dihydrochloride (9CI), 3-[[2-[(2-Chloroethyl)amino]ethyl]amino]propyl dihydrogen phosphate, dihydrochloride |
| InChI formel | InChI=1S/C7H18ClN2O4P.2ClH/c8-2-4-10-6-5-9-3-1-7-14-15(11,12)13;;/h9-10H,1-7H2,(H2,11,12,13);2*1H |
| LEDER | Cl.Cl.OP(=O)(O)OCCCNCCNCCCl |
| IUPAC-namn | 3-[2-(2-chloroethylamino)ethylamino]propyl dihydrogen phosphate;dihydrochloride |
| Molekylvikt (g/mol) | 333.58 |
| Visa mer |