Läs mer
3-(2,2-Dichlorovinyl)-2,2-dimethylcyclopropanecarbonyl Chloride, TRC
Produktkod. 30378712
Change view
Kvantitet:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30378712 250 mg
30384918 1 g
30381526 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30378712 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD435678250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 52314-67-7
Molekylformel C8H9Cl3O
Molekylvikt (g/mol) 227.52
Synonym 3-(2,2-Dichloroethenyl)-2,2-dimethylcyclopropanecarbonyl Chloride, 2,2-Dimethyl-3-(2,2-dichloroethenyl)cyclopropanecarbonyl Chloride, 2,2-Dimethyl-3-(2,2-dichlorovinyl)cyclopropanecarbonyl Chloride, 3-(2,2-Dichlorovinyl)-2,2-dimethyl-1-cyclopropanecarbonyl Chloride
IUPAC-namn 3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carbonyl chloride
LEDER CC1(C)C(C=C(Cl)Cl)C1C(=O)Cl
Specifikationer
Kemiskt namn eller material 3-(2,2-Dichlorovinyl)-2,2-dimethylcyclopropanecarbonyl Chloride
Kvantitet 250 mg
InChI formel InChI=1S/C8H9Cl3O/c1-8(2)4(3-5(9)10)6(8)7(11)12/h3-4,6H,1-2H3
Formel vikt 225.9719