Läs mer
3-(2,4,6-Trimethylphenylsulfonyl)-1,2,4-triazole, TRC
Produktkod. 30455672
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30455672 500 mg
30452143 1 g
30451608 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30455672 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT482395500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 149591-20-8
Molekylformel C11 H13 N3 O2 S
Molekylvikt (g/mol) 251.3
Synonym 1H-1,2,4-Triazole, 3-[(2,4,6-trimethylphenyl)sulfonyl]- (9CI), 5-[(2,4,6-Trimethylphenyl)sulfonyl]-1H-1,2,4-triazole, 3-(2,4,6-Trimethylbenzenesulfonyl)-1,2,4-triazole, 3-(2,4,6-Trimethylphenylsulfonyl)-1,2,4-triazole
IUPAC-namn 3-(2,4,6-trimethylphenyl)sulfonyl-4H-1,2,4-triazole
LEDER Cc1cc(C)c(c(C)c1)S(=O)(=O)c2nnc[nH]2
Specifikationer
Kemiskt namn eller material 3-(2,4,6-Trimethylphenylsulfonyl)-1,2,4-triazole
Kvantitet 500 mg
InChI formel InChI=1S/C11H13N3O2S/c1-7-4-8(2)10(9(3)5-7)17(15,16)11-12-6-13-14-11/h4-6H,1-3H3,(H,12,13,14)
Formel vikt 251.0728