Läs mer
3-[2-[4-(Diphenylmethyl)-1-piperazinyl]ethyl] 5-Methyl 2,6-Dimethyl-4-(3-nitrophenyl)-pyridine-3,5-dicarboxylate Dihydrochloride, TRC
Produktkod. 30364565
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30364565 250 mg
1 options
Produktkod. 30364565 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD222760250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-[2-[4-(Diphenylmethyl)-1-piperazinyl]ethyl] 5-Methyl 2,6-Dimethyl-4-(3-nitrophenyl)-pyridine-3,5-dicarboxylate Dihydrochloride
CAS 2227064-54-0
Molekylformel C35H36N4O6.2HCl
Kvantitet 250 mg
Synonym O3-[2-(4-Benzhydrylpiperazin-1-yl)ethyl] O5-methyl 2,6-dimethyl-4-(3-nitrophenyl)pyridine-3,5-dicarboxylate Dihydrochloride, 3,5-Pyridinedicarboxylic acid, 2,6-dimethyl-4-(3-nitrophenyl)-, 3-[2-[4-(diphenylmethyl)-1-piperazinyl]ethyl] 5-methyl ester, hydrochloride (1:2), 3-[2-[4-(Diphenylmethyl)-1-piperazinyl]ethyl] 5-Methyl 2,6-Dimethyl-4-(3-nitrophenyl)-pyridine-3,5-dicarboxylate Dihydrochloride
InChI formel InChI=1S/C34H34N4O6.2ClH/c1-24-30(31(29(23-35-24)33(39)43-2)27-14-9-15-28(22-27)38(41)42)34(40)44-21-20-36-16-18-37(19-17-36)32(25-10-5-3-6-11-25)26-12-7-4-8-13-26;;/h3-15,22-23,32H,16-21H2,1-2H3;2*1H
LEDER O=C(OC)C1=CN=C(C(C(OCCN2CCN(CC2)C(C3=CC=CC=C3)C4=CC=CC=C4)=O)=C1C5=CC(N(=O)=O)=CC=C5)C.[H]Cl.[H]Cl
IUPAC-namn 3-O-[2-(4-benzhydrylpiperazin-1-yl)ethyl] 5-O-methyl 2-methyl-4-(3-nitrophenyl)pyridine-3,5-dicarboxylate;dihydrochloride
Molekylvikt (g/mol) 681.61