missing translation for 'onlineSavingsMsg'
Learn More

3(2),6,6-Trimethylfulvene, mixture of isomers, tech. 90%, Thermo Scientific™

Catalog Number p-7026049
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
30953-08-3
C6H12
MFCD02683073
This item is not returnable. View return policy
30953-08-3
C6H12
MFCD02683073

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN3295
Formula Weight 120.19
Percent Purity 90%
Grade Technical
Chemical Name or Material 3(2),6,6-Trimethylfulvene, mixture of isomers
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'