missing translation for 'onlineSavingsMsg'
Learn More

3-(2-bromo-5-methoxyphenyl)-3-oxopropanenitrile, Thermo Scientific™

Catalog Number p-4391215
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
175136-65-9
C10H8BrNO2
254.083
NVZISNDFOXXQSQ-UHFFFAOYSA-N
3-2-bromo-5-methoxyphenyl-3-oxopropanenitrile, maybridge1_002847, 3-2-bromo-5-methoxy-phenyl-3-oxo-propanenitrile
2804251
3-(2-bromo-5-methoxyphenyl)-3-oxopropanenitrile
COC1=CC(=C(C=C1)Br)C(=O)CC#N
This item is not returnable. View return policy
175136-65-9
C10H8BrNO2
254.083
NVZISNDFOXXQSQ-UHFFFAOYSA-N
3-2-bromo-5-methoxyphenyl-3-oxopropanenitrile, maybridge1_002847, 3-2-bromo-5-methoxy-phenyl-3-oxo-propanenitrile
2804251
3-(2-bromo-5-methoxyphenyl)-3-oxopropanenitrile
COC1=CC(=C(C=C1)Br)C(=O)CC#N
Assay Percent Range 97%
Quantity 1 g
Formula Weight 254.08
Percent Purity 97%
Chemical Name or Material 3-(2-Bromo-5-methoxyphenyl)-3-oxopropanenitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'