Läs mer
3-(2-Chlorobenzoyl)-4-hydroxy-2H-1-benzopyran-2-one, TRC
Produktkod. 30349366
Change view
Kvantitet:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30349366 50 mg
30363899 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30349366 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC46341550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-(2-Chlorobenzoyl)-4-hydroxy-2H-1-benzopyran-2-one
CAS 181938-56-7
Molekylformel C16H9ClO4
Kvantitet 50 mg
Synonym NSC 133803
InChI formel InChI=1S/C16H9ClO4/c17-11-7-3-1-5-9(11)14(18)13-15(19)10-6-2-4-8-12(10)21-16(13)20/h1-8,19H
LEDER OC1=C(C(=O)c2ccccc2Cl)C(=O)Oc3ccccc13
IUPAC-namn 3-(2-chlorobenzoyl)-4-hydroxychromen-2-one
Molekylvikt (g/mol) 300.69
Formel vikt 300.0189