Läs mer
3-(2-Chloroethyl)-9-hydroxy-2-methyl-4H-Pyrido[1,2-a]pyrimidin-4-one Hydrochloride, TRC
Produktkod. 30367250
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30367250 25 mg
1 options
Produktkod. 30367250 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36424025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-(2-Chloroethyl)-9-hydroxy-2-methyl-4H-Pyrido[1,2-a]pyrimidin-4-one Hydrochloride
CAS 849727-62-4
Molekylformel C11 H11 Cl N2 O2 . Cl H
Kvantitet 25 mg
Synonym 4H-Pyrido[1,2-a]pyrimidin-4-one, 3-(2-chloroethyl)-9-hydroxy-2-methyl-, hydrochloride (1:1), 4H-Pyrido[1,2-a]pyrimidin-4-one, 3-(2-chloroethyl)-9-hydroxy-2-methyl-, monohydrochloride (9CI), 3-(2-Chloroethyl)-9-hydroxy-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one hydrochloride, 9-Hydroxy-3-(2-chloroethyl)-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one hydrochloride
InChI formel InChI=1S/C11H11ClN2O2.ClH/c1-7-8(4-5-12)11(16)14-6-2-3-9(15)10(14)13-7;/h2-3,6,15H,4-5H2,1H3;1H
LEDER Cl.CC1=C(CCCl)C(=O)N2C=CC=C(O)C2=N1
IUPAC-namn 3-(2-chloroethyl)-9-hydroxy-2-methylpyrido[1,2-a]pyrimidin-4-one;hydrochloride
Molekylvikt (g/mol) 275.13
Formel vikt 274.0276