Läs mer
3-((2-Chloroethyl)amino)propyl Dihydrogen Phosphate, TRC
Produktkod. 30355841
Change view
Kvantitet:
10 mg
20 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30355841 50 mg
30362559 10 mg
30350507 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30355841 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC29333050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 22608-58-8
Molekylformel C5 H13 Cl N O4 P
Molekylvikt (g/mol) 217.59
Synonym 1-Propanol, 3-[(2-chloroethyl)amino]-, dihydrogen phosphate (ester) (8CI,9CI), Ifosfamide Imp. A
IUPAC-namn 3-(2-chloroethylamino)propyl dihydrogen phosphate
LEDER OP(=O)(O)OCCCNCCCl
Specifikationer
Kemiskt namn eller material 3-[(2-Chloroethyl)amino]propyl Dihydrogen Phosphate
Kvantitet 50 mg
InChI formel InChI=1S/C5H13ClNO4P/c6-2-4-7-3-1-5-11-12(8,9)10/h7H,1-5H2,(H2,8,9,10)
Formel vikt 217.0271