missing translation for 'onlineSavingsMsg'
Läs mer

3-(2-Chloroethynyl)-2,2-dimethylcyclopropanecarboxylic Acid, TRC

Produktkod. 30354558
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30354558 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30354558 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC630080500MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 3-(2-Chloroethynyl)-2,2-dimethylcyclopropanecarboxylic Acid
CAS 63406-22-4
Molekylformel C8H9ClO2
Kvantitet 500 mg
Synonym 2,2-Dimethyl-3-chloroethynylcyclopropanecarboxylic Acid
InChI formel InChI=1S/C8H9ClO2/c1-8(2)5(3-4-9)6(8)7(10)11/h5-6H,1-2H3,(H,10,11)
LEDER ClC#CC1C(C(O)=O)C1(C)C
IUPAC-namn 3-(2-chloroethynyl)-2,2-dimethylcyclopropane-1-carboxylic acid
Molekylvikt (g/mol) 172.61
Formel vikt 172.0291
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.