Läs mer
3-(2-Chloropropionyl)amino-4-methyl-2-methoxycarbonylthiophene, TRC
Produktkod. 30351021
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30351021 5 mg
30367104 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30351021 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3808735MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3-(2-Chloropropionyl)amino-4-methyl-2-methoxycarbonylthiophene
CAS 23964-56-9
Molekylformel C10H12ClNO3S
Kvantitet 5 mg
Synonym Methyl 3-(2-chloropropanoylamino)-4-methyl-2-thiophenecarboxylate
InChI formel InChI=1S/C10H12ClNO3S/c1-5-4-16-8(10(14)15-3)7(5)12-9(13)6(2)11/h4,6H,1-3H3,(H,12,13)
LEDER O=C(OC)C1=C(NC(C(C)Cl)=O)C(C)=CS1
IUPAC-namn methyl 3-(2-chloropropanamido)-4-methylthiophene-2-carboxylate
Molekylvikt (g/mol) 261.73
Formel vikt 261.2