Läs mer
3-(2-Deoxy-Beta-D-erythro-pentofuranosyl)-3,5-dihydropyrimido[1,2-a]purine-6,10-dione, TRC
Produktkod. 30362322
Change view
Kvantitet:
1 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30362322 25 mg
30358692 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30362322 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23518525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 895571-79-6
Molekylformel C13 H13 N5 O5
Molekylvikt (g/mol) 319.273
IUPAC-namn 3-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5H-pyrimido[1,2-a]purine-6,10-dione
LEDER OC[C@H]1O[C@H](C[C@@H]1O)n2cnc3C(=O)N4C=CC(=O)NC4=Nc23
Specifikationer
Kemiskt namn eller material 3-(2-Deoxy-b-D-erythro-pentofuranosyl)-3,5-dihydropyrimido[1,2-a]purine-6,10-dione
Kvantitet 25 mg
InChI formel InChI=1S/C13H13N5O5/c19-4-7-6(20)3-9(23-7)18-5-14-10-11(18)16-13-15-8(21)1-2-17(13)12(10)22/h1-2,5-7,9,19-20H,3-4H2,(H,15,16,21)/t6-,7+,9+/m0/s1
Formel vikt 319.092