Läs mer
3-(2-Deoxy-Beta-D-erythro-pentofuranosyl)-4,6,7,8-tetrahydro-8-hydroxypyrimido[1,2-a]purin-10(3H)-one (~90%), TRC
Produktkod. 30366471
Change view
Kvantitet:
0.5 mg
2.5 mg
10 mg
Förpackningsstorlek:
0.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30366471 2.5 mg -
30359237 0.5 mg 0.5mg
30362156 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30366471 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2825302.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 91021-93-1
Molekylformel C13H17N5O5
Molekylvikt (g/mol) 323.3
Synonym gamma-Hydroxy-1,N2-propano-2'-deoxyguanosine
LEDER OC[C@H]1O[C@H](C[C@@H]1O)n2cnc3C(=O)N4C(O)CCN=C4Nc23
Specifikationer
Kemiskt namn eller material 3-(2-Deoxy-beta-D-erythro-pentofuranosyl)-4,6,7,8-tetrahydro-8-hydroxypyrimido[1,2-a]purin-10(3H)-one
Kvantitet 2.5 mg
InChI formel InChI=1S/C13H17N5O5/c19-4-7-6(20)3-9(23-7)17-5-15-10-11(17)16-13-14-2-1-8(21)18(13)12(10)22/h5-9,19-21H,1-4H2,(H,14,16)/t6-,7+,8?,9+/m0/s1
Formel vikt 323.123