Läs mer
3-[[[2-[(Diaminomethylene)amino]-4-thiazolyl]methyl]thio]propanimidamide Hydrochloride, TRC
Produktkod. 30353978
Change view
Kvantitet:
10 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30353978 10 mg
30362684 250 mg
30362449 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30353978 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD41767010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 76833-47-1
Molekylformel C8 H14 N6 S2 . Cl H
Molekylvikt (g/mol) 294.83
Synonym Propanimidamide, 3-[[[2-[(aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]-, hydrochloride (1:1), Propanimidamide, 3-[[[2-[(aminoiminomethyl)amino]-4-thiazolyl]methyl]thio]-, monohydrochloride (9CI), 3-[[[2-[(Diaminomethylene)amino]thiazol-4-yl]-methyl]sulfanyl]propanimidamide Hydrochloride, Famotidine Imp. A (EP) as Hydrochloride
IUPAC-namn 3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]propanimidamide;hydrochloride
LEDER Cl.NC(=Nc1nc(CSCCC(=N)N)cs1)N
Specifikationer
Kemiskt namn eller material 3-[([2-[(Diaminomethylene)amino]thiazol-4-yl]-methyl)sulfanyl]propanimidamideHydrochloride
Kvantitet 10 mg
InChI formel InChI=1S/C8H14N6S2.ClH/c9-6(10)1-2-15-3-5-4-16-8(13-5)14-7(11)12;/h4H,1-3H2,(H3,9,10)(H4,11,12,13,14);1H
Formel vikt 294.0488