Learn More
3-[2-(Diaminomethyleneamino)-1,3-thiazol-4-ylmethylsulphinyl]-N-sulphamoylpropanamide (Famotidine Metabolite), TRC
Product Code. 30363034
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity Unit Size
30363034 25 mg 25mg
1 options
Product Code. 30363034 Supplier Toronto Research Chemicals Supplier No. TRCD41636725MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-[2-(Diaminomethyleneamino)-1,3-thiazol-4-ylmethylsulfinyl]-N-sulfamoylpropanamide
CAS 1020719-36-1
Purity Grade Notes HPLC
Molecular Formula C8 H14 N6 O4 S3
Quantity 25 mg
Synonym 3-[[[2-[(Diaminomethylene)-amino]thiazol-4-yl]methyl]sulfinyl]-N-sulfamoyl-propanamide, Famotidine Imp. I (Pharmeuropa)
InChI Formula InChI=1S/C8H14N6O4S3/c9-7(10)13-8-12-5(3-19-8)4-20(16)2-1-6(15)14-21(11,17)18/h3H,1-2,4H2,(H,14,15)(H2,11,17,18)(H4,9,10,12,13)
SMILES NC(=Nc1nc(CS(=O)CCC(=O)NS(=O)(=O)N)cs1)N
IUPAC Name 3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfinyl]-N-sulfamoylpropanamide
Molecular Weight (g/mol) 354.43
Formula Weight 354.0239
Percent Purity >95