missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Isopropylphenoxy)piperidine, Thermo Scientific™

Catalog Number p-7033821
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
902837-35-8
C14H21NO
219.328
MFCD08061106
DFJMRKMGDKDNAX-UHFFFAOYSA-N
3-2-isopropylphenoxy piperidine, 3-2-propan-2-yl phenoxy piperidine
44721294
3-(2-propan-2-ylphenoxy)piperidine
CC(C)C1=CC=CC=C1OC2CCCNC2
This item is not returnable. View return policy
902837-35-8
C14H21NO
219.328
MFCD08061106
DFJMRKMGDKDNAX-UHFFFAOYSA-N
3-2-isopropylphenoxy piperidine, 3-2-propan-2-yl phenoxy piperidine
44721294
3-(2-propan-2-ylphenoxy)piperidine
CC(C)C1=CC=CC=C1OC2CCCNC2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 219.33
Chemical Name or Material 3-(2-Isopropylphenoxy)piperidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'