missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Thienyl)pyridine, 99%, Thermo Scientific™

Catalog Number 11329395
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
21298-53-3
C9H7NS
161.222
MFCD01860052
ZHBLIWDUZHFSJW-UHFFFAOYSA-N
3-2-thienyl pyridine, 3-thiophen-2-yl pyridine, 2-3-pyridyl thiophene, 3-thiophen-2-yl-pyridine, pyridine,3-2-thienyl, 3-thien-2-ylpyridine, acmc-1clkc, 3-2-thienyl-pyridine, 3-thien-2-yl pyridine, 3-thiophene-2-yl-pyridine
595145
3-thiophen-2-ylpyridine
C1=CC(=CN=C1)C2=CC=CS2
This item is not returnable. View return policy
21298-53-3
C9H7NS
161.222
MFCD01860052
ZHBLIWDUZHFSJW-UHFFFAOYSA-N
3-2-thienyl pyridine, 3-thiophen-2-yl pyridine, 2-3-pyridyl thiophene, 3-thiophen-2-yl-pyridine, pyridine,3-2-thienyl, 3-thien-2-ylpyridine, acmc-1clkc, 3-2-thienyl-pyridine, 3-thien-2-yl pyridine, 3-thiophene-2-yl-pyridine
595145
3-thiophen-2-ylpyridine
C1=CC(=CN=C1)C2=CC=CS2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Boiling Point 95°C to 100°C (1 mmHg)
Refractive Index 1.648
Quantity 250 mg
Beilstein 971159
Formula Weight 161.23
Percent Purity 99%
Chemical Name or Material 3-(2-Thienyl)pyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'