Läs mer
3-[[(2S,3R)-2-[(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)morpholino]methyl]-1H-1,2,4-triazol-5(4H)-one ((S,R,S)-Aprepitant), Mikromol™
Produktkod. 30052307
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30052307 100 mg
30049297 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30052307 Leverantör Mikromol Leverantörsnummer MM3251.02
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-[[(2S,3R)-2-[(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)morpholino]methyl]-1H-1,2,4-triazol-5(4H)-one ((S,R,S)-Aprepitant)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S,R,S)-Aprepitant
CAS 172822-29-6
Analyt- eller komponentnamn (S,R,S)-Aprepitant
Frakt skick Room Temperature
Molekylformel C23 H21 F7 N4 O3
Certifikat och efterlevnad ISO 17025
Synonym 3H-1,2,4-Triazol-3-one, 5-[[(2S,3R)-2-[(1S)-1-[3,5-bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)-4-morpholinyl]methyl]-1,2-dihydro-, ent-Aprepitant, Aprepitant Related Compound B (USP), 3-[[(2S,3R)-2-[(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)morpholino]methyl]-1H-1,2,4-triazol-5(4H)-one, 3H-1,2,4-Triazol-3-one, 5-[[2-[1-[3,5-bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)-4-morpholinyl]methyl]-1,2-dihydro-, [2S-[2alpha(R*),3alpha]]-, Aprepitant Related Compound B
InChI formel InChI=1S/C23H21F7N4O3/c1-12(14-8-15(22(25,26)27)10-16(9-14)23(28,29)30)37-20-19(13-2-4-17(24)5-3-13)34(6-7-36-20)11-18-31-21(35)33-32-18/h2-5,8-10,12,19-20H,6-7,11H2,1H3,(H2,31,32,33,35)/t12-,19+,20-/m0/s1
LEDER C[C@H](O[C@@H]1OCCN(CC2=NNC(=O)N2)[C@@H]1c3ccc(F)cc3)c4cc(cc(c4)C(F)(F)F)C(F)(F)F
IUPAC-namn 3-[[(2S,3R)-2-[(1S)-1-[3,5-bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)morpholin-4-yl]methyl]-1,4-dihydro-1,2,4-triazol-5-one
Molekylvikt (g/mol) 534.43
Kvantitet 100 mg
Formel vikt 534.1502 g/mol