Läs mer
3-[3-(2,3-Dihydro-2-oxo-1H-benzimidazol-1-yl)propyl] Domperidone (Domperidone Impurity D), TRC
Produktkod. 30010571
Change view
Kvantitet:
2.5 mg
10 mg
100 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30010571 10 mg -
30005985 2.5 mg 2.5mg
17928124 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010571 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44945510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1614255-34-3
Molekylformel C32 H34 Cl N7 O3
Molekylvikt (g/mol) 600.11
Synonym 2H-Benzimidazol-2-one, 5-chloro-3-[3-(2,3-dihydro-2-oxo-1H-benzimidazol-1-yl)propyl]-1-[1-[3-(2,3-dihydro-2-oxo-1H-benzimidazol-1-yl)propyl]-4-piperidinyl]-1,3-dihydro-, 5-Chloro-3-[3-(2,3-dihydro-2-oxo-1H-benzimidazol-1-yl)propyl]-1-[1-[3-(2,3-dihydro-2-oxo-1H-benzimidazol-1-yl)propyl]-4-piperidinyl]-1,3-dihydro-2H-benzimidazol-2-one, 5-Chloro-3-[3-(2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)-propyl]-1-[1-[3-(2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)propyl]piperidin-4-yl]-1,3-dihydro-2H-benzimidazol-2-one
IUPAC-namn 5-chloro-3-[3-(2-oxo-3H-benzimidazol-1-yl)propyl]-1-[1-[3-(2-oxo-3H-benzimidazol-1-yl)propyl]piperidin-4-yl]benzimidazol-2-one
LEDER Clc1ccc2N(C3CCN(CCCN4C(=O)Nc5ccccc45)CC3)C(=O)N(CCCN6C(=O)Nc7ccccc67)c2c1
Specifikationer
Kemiskt namn eller material 3-[3-(2,3-Dihydro-2-oxo-1H-benzimidazol-1-yl)propyl] Domperidone
Kvantitet 10 mg
InChI formel InChI=1S/C32H34ClN7O3/c33-22-11-12-28-29(21-22)39(18-6-17-38-27-10-4-2-8-25(27)35-31(38)42)32(43)40(28)23-13-19-36(20-14-23)15-5-16-37-26-9-3-1-7-24(26)34-30(37)41/h1-4,7-12,21,23H,5-6,13-20H2,(H,34,41)(H,35,42)
Formel vikt 599.2412