Läs mer
3'-[3-(3-Azido-2,3-dideoxy-b-D-erythro-pentofuranosyl)-3,6-dihydro-5-methyl-2,6-dioxo-1(2H)-pyrimidinyl]-3'-deoxy-thymidine, TRC
Produktkod. 30334598
Change view
Kvantitet:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30334598 5 mg
30343697 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30334598 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA8250055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3'-[3-(3-Azido-2,3-dideoxy-Beta-D-erythro-pentofuranosyl)-3,6-dihydro-5-methyl-2,6-dioxo-1(2H)-pyrimidinyl]-3'-deoxy-thymidine
CAS 148665-49-0
Molekylformel C20 H25 N7 O8
Kvantitet 5 mg
Synonym Thymidine, 3'-[3-(3-azido-2,3-dideoxy-β-D-erythro-pentofuranosyl)-3,6-dihydro-5-methyl-2,6-dioxo-1(2H)-pyrimidinyl]-3'-deoxy-, 3'-[3-(3-Azido-2,3-dideoxy-β-D-erythro-pentofuranosyl)-3,6-dihydro-5-methyl-2,6-dioxo-1(2H)-pyrimidinyl]-3'-deoxythymidine, Zidovudine Imp. G (EP), 1-[(2R,4S,5S)-4-azido-5-(hydroxymethyl)tetrahydrofuran-2-yl]-3-[(2S,3S,5R)-2-(hydroxymethyl)-5-(5-methyl-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)tetrahydrofuran-3-yl]-5-methylpyrimidine-2,4(1H,3H)-dione
InChI formel InChI=1S/C20H25N7O8/c1-9-5-25(19(32)22-17(9)30)16-4-12(14(8-29)35-16)27-18(31)10(2)6-26(20(27)33)15-3-11(23-24-21)13(7-28)34-15/h5-6,11-16,28-29H,3-4,7-8H2,1-2H3,(H,22,30,32)/t11-,12-,13+,14+,15+,16+/m0/s1
LEDER CC1=CN([C@H]2C[C@@H]([C@@H](CO)O2)N3C(=O)N(C=C(C)C3=O)[C@H]4C[C@H](N=[N+]=[N-])[C@@H](CO)O4)C(=O)NC1=O
IUPAC-namn 1-[(2R,4S,5S)-4-azido-5-(hydroxymethyl)oxolan-2-yl]-3-[(2S,3S,5R)-2-(hydroxymethyl)-5-(5-methyl-2,4-dioxopyrimidin-1-yl)oxolan-3-yl]-5-methylpyrimidine-2,4-dione
Molekylvikt (g/mol) 491.45
Formel vikt 491.1765