Läs mer
3,3,3-Trideuterio-2,2-difluoro-propanoic Acid, TRC
Produktkod. 30367241
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30367241 50 mg
30354641 100 mg
30361956 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30367241 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD22310050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1391033-19-4
Molekylformel C3 D3 H F2 O2
Molekylvikt (g/mol) 113.078
IUPAC-namn 3,3,3-trideuterio-2,2-difluoropropanoic acid
LEDER [2H]C([2H])([2H])C(F)(F)C(=O)O
Specifikationer
Kemiskt namn eller material 3,3,3-Trideuterio-2,2-difluoro-propanoic Acid
Kvantitet 50 mg
InChI formel InChI=1S/C3H4F2O2/c1-3(4,5)2(6)7/h1H3,(H,6,7)/i1D3
Formel vikt 113.037