missing translation for 'onlineSavingsMsg'
Learn More

3,3',4,4'-Benzophenonetetracarboxylic dianhydride, 97+%, Thermo Scientific Chemicals

Catalog Number p-7023208
Click to view available options
Quantity:
100 g
500 g
2.5 kg
This item is not returnable. View return policy
2421-28-5
C17H6O7
322.228
MFCD00005923
VQVIHDPBMFABCQ-UHFFFAOYSA-N
3,3',4,4'-benzophenonetetracarboxylic dianhydride, benzophenonetetracarboxylic dianhydride, benzophenonetetracarboxylic acid dianhydride, 1,3-isobenzofurandione, 5,5'-carbonylbis, 4,4'-carbonylbis phthalic anhydride, 4,4'-carbonyldiphthalic anhydride, 4,4'-diphthalic anhydride ketone, benzophenonetetracarboxylic anhydride, phthalic anhydride, 4,4'-carbonyldi, unii-y61gva8097
75498
5-(1,3-dioxo-2-benzofuran-5-carbonyl)-2-benzofuran-1,3-dione
C1=CC2=C(C=C1C(=O)C3=CC4=C(C=C3)C(=O)OC4=O)C(=O)OC2=O
This item is not returnable. View return policy
2421-28-5
C17H6O7
322.228
MFCD00005923
VQVIHDPBMFABCQ-UHFFFAOYSA-N
3,3',4,4'-benzophenonetetracarboxylic dianhydride, benzophenonetetracarboxylic dianhydride, benzophenonetetracarboxylic acid dianhydride, 1,3-isobenzofurandione, 5,5'-carbonylbis, 4,4'-carbonylbis phthalic anhydride, 4,4'-carbonyldiphthalic anhydride, 4,4'-diphthalic anhydride ketone, benzophenonetetracarboxylic anhydride, phthalic anhydride, 4,4'-carbonyldi, unii-y61gva8097
75498
5-(1,3-dioxo-2-benzofuran-5-carbonyl)-2-benzofuran-1,3-dione
C1=CC2=C(C=C1C(=O)C3=CC4=C(C=C3)C(=O)OC4=O)C(=O)OC2=O

BTDA polymer fine free-flowing powder suitable for general purposes. BTDA Microfine Micronized powder facilitates mixing in epoxy resins. BTDA ultrapure high-purity, high-assay powder for critical polyimide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
BTDA polymer fine free-flowing powder suitable for general purposes. BTDA Microfine Micronized powder facilitates mixing in epoxy resins. BTDA ultrapure high-purity, high-assay powder for critical polyimide synthesis.

Solubility
Soluble in DMF (0.1 g/mL). Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 220°C to 226°C
Density 1.57
Boiling Point 320°C (5 mmHg)
Flash Point 324°C (615°F)
Quantity 100 g
Beilstein 761777
Sensitivity Moisture sensitive
Solubility Information Soluble in DMF (0.1g/mL). Reacts with water.
Formula Weight 322.23
Percent Purity ≥97%
Chemical Name or Material 3,3',4,4'-Benzophenonetetracarboxylic dianhydride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'